Learn More
ACROBiosystems Recombinant Protein;Human Ephrin-A4 / EFNA4 Protein;HEK293;100UG;EA4,EFNA4,EPLG4,LERK4,LERK-4,EFNA-4,EPLG-4,Ephrin-A4
Supplier:  ACROBiosystems EA4H5229100UG
Encompass_Preferred
Recombinant Protein;Human Ephrin-A4 / EFNA4 Protein;100UG;Human Ephrin-A4, His Tag (EA4-H5229) is expressed from human 293 cells (HEK293). It contains AA Leu 26 - Gly 171 (Accession # AAI07484).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 17.2 kDa. The protein migrates as 20-22 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8700