Learn More
Aviva Systems Biology Corp EPHX1 Antibody - middle region (ARP54286_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54286_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against EPHX1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human EPHX1. Alternate names: EPHX, EPOX, MEH, HYL1ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-61158