Learn More
Medchemexpress LLC EPHX1 Human His-SU 20ug
SDP
Catalog No. 5000183372
Encompass_Preferred
Recombinant human EPHX1 (epoxide hydrolase 1) is provided as a purified, N-terminal His-SUMO-tagged protein expressed in E. coli for use in biochemical characterization, enzyme assays, and lipid metabolism research. It catalyzes hydrolysis of aromatic and aliphatic epoxides and supports studies of xenobiotic biotransformation and endogenous lipid signaling.
  • N-terminal His-SUMO tag for efficient purification and improved solubility.
  • High purity (>90%) by reducing SDS-PAGE for reliable assay performance.
  • Approximately 66 kDa molecular weight for size reference in analyses.
  • Low endotoxin level (<1 EU/μg) suitable for sensitive biochemical assays.
  • Available in small quantities (20 μg, 50 μg, 100 μg) for screening and pilot studies.
Catalog No. 50-001-83372 Supplier Medchemexpress LLC Supplier No. HYP7218420UG
Recombinant human EPHX1 (epoxide hydrolase 1) is provided as a purified, N-terminal His-SUMO-tagged protein expressed in E. coli for use in biochemical characterization, enzyme assays, and lipid metabolism research. It catalyzes hydrolysis of aromatic and aliphatic epoxides and supports studies of xenobiotic biotransformation and endogenous lipid signaling.
  • N-terminal His-SUMO tag for efficient purification and improved solubility.
  • High purity (>90%) by reducing SDS-PAGE for reliable assay performance.
  • Approximately 66 kDa molecular weight for size reference in analyses.
  • Low endotoxin level (<1 EU/μg) suitable for sensitive biochemical assays.
  • Available in small quantities (20 μg, 50 μg, 100 μg) for screening and pilot studies.