Learn More
Selleck Chemical LLC EPI-001 10mg
SDP
Supplier:  Selleck Chemical LLC S795510MG
Encompass_Preferred
EPI-001 is an androgen receptor N-terminal domain antagonist with IC50 of ?6 ?M and a selective PPAR-gamma modulator. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3329