Learn More
Medchemexpress LLC EPI-743 (α-Tocotrienol quinone) | 1213269-98-7 | 98.1% | 440.66 | 5 MG
SDP
Catalog No. 5000390741
Encompass_Preferred
EPI-743 (α-Tocotrienol quinone), also known as Vatiquinone, is a potent cellular oxidative stress protectant and ferroptosis inhibitor. It is a synthetic analog of vitamin E, used in the study of mitochondrial diseases, and targets the repletion of reduced intracellular glutathione.
  • Potent cellular oxidative stress protectant
  • Inhibits ferroptosis in cells
  • Used for the study of mitochondrial diseases
  • Synthetic analog of vitamin E
  • Targets the repletion of reduced intracellular glutathione
  • Oral activity
  • Prevents RSL3-induced ferroptosis in PCH6 patient fibroblasts
  • Prevents BODIPY 581/591 C11 lipid oxidation
  • Inhibits RSL3-mediated ferroptotic cell death in B-lymphocytes
Catalog No. 50-003-90741 Supplier Medchemexpress LLC Supplier No. HY167725MG
EPI-743 (α-Tocotrienol quinone), also known as Vatiquinone, is a potent cellular oxidative stress protectant and ferroptosis inhibitor. It is a synthetic analog of vitamin E, used in the study of mitochondrial diseases, and targets the repletion of reduced intracellular glutathione.
  • Potent cellular oxidative stress protectant
  • Inhibits ferroptosis in cells
  • Used for the study of mitochondrial diseases
  • Synthetic analog of vitamin E
  • Targets the repletion of reduced intracellular glutathione
  • Oral activity
  • Prevents RSL3-induced ferroptosis in PCH6 patient fibroblasts
  • Prevents BODIPY 581/591 C11 lipid oxidation
  • Inhibits RSL3-mediated ferroptotic cell death in B-lymphocytes