Learn More
Medchemexpress LLC EPI-743 (α-Tocotrienol quinone) | 1213269-98-7 | 98.1% | 440.66 | 50 MG
SDP
Catalog No. 5000391262
Encompass_Preferred
EPI-743 (Vatiquinone; α-Tocotrienol quinone; PTC-743; NCT04378075) is a potent cellular oxidative stress protectant that inhibits ferroptosis in cells. It can be used for the study of mitochondrial diseases. EPI-743 is a synthetic analog of vitamin E with oral activity, targeting the repletion of reduced intracellular glutathione.
  • Potent cellular oxidative stress protectant
  • Inhibits ferroptosis in cells
  • Used for the study of mitochondrial diseases
  • Synthetic analog of vitamin E
  • Targets the repletion of reduced intracellular glutathione
Catalog No. 50-003-91262 Supplier Medchemexpress LLC Supplier No. HY1677250MG
EPI-743 (Vatiquinone; α-Tocotrienol quinone; PTC-743; NCT04378075) is a potent cellular oxidative stress protectant that inhibits ferroptosis in cells. It can be used for the study of mitochondrial diseases. EPI-743 is a synthetic analog of vitamin E with oral activity, targeting the repletion of reduced intracellular glutathione.
  • Potent cellular oxidative stress protectant
  • Inhibits ferroptosis in cells
  • Used for the study of mitochondrial diseases
  • Synthetic analog of vitamin E
  • Targets the repletion of reduced intracellular glutathione