Learn More
AdipoGen epi-Aszonalenin A (1 mg)
SDP
Supplier:  AdipoGen AGCN20163M001
Encompass_Preferred
Chemical. CAS: 908853-14-5. Formula: C25H25N3O3. MW: 415.4. Isolated from Neosartorya fischeri. Derivative of aszonalenin, a substance P inhibitor.
Catalog No. 50-205-3551