Learn More
Cayman Chemical epi-Ivermectin B1a, Purity: ≥95%, 1 mg
SDP
Catalog No. 502934667
Encompass_Preferred
An ivermectin B1a degradation product; less active than ivermectin against T. urticae (LC90s = 4.0 and 0.038 ppm, respectively)
Catalog No. 50-293-4667 Supplier Cayman Chemical Supplier No. 252111 MG
An ivermectin B1a degradation product; less active than ivermectin against T. urticae (LC90s = 4.0 and 0.038 ppm, respectively)