Learn More
Cayman Chemical epI-IvermectIn B1a 5mg
SDP
Catalog No. 502934617
Encompass_Preferred
An ivermectin B1a degradation product; less active than ivermectin against T. urticae (LC90s = 4.0 and 0.038 ppm, respectively)
Catalog No. 50-293-4617 Supplier Cayman Chemical Supplier No. 252115 MG
An ivermectin B1a degradation product; less active than ivermectin against T. urticae (LC90s = 4.0 and 0.038 ppm, respectively)