Learn More
Epigentek Inc EpiQuik MeDIP Ultra Kit (24 reactions)
SDP
Catalog No. NC1033408
Encompass_Preferred
For antibody-based capture of methylated DNA fragments for microarray, PCR, or NGS applications
Catalog No. NC1033408 Supplier Epigentek Inc Supplier No. P105224
For antibody-based capture of methylated DNA fragments for microarray, PCR, or NGS applications