Learn More
Cayman Chemical Epiallopregnanolone, 567-01-1, 1 mg
SDP
Supplier:  Cayman Chemical 184311
Encompass_Preferred
A metabolite of progesterone and an inactive isomer of allopregnanolone, a positive modulator of GABAA receptors
Catalog No. NC1448324