Learn More
Enzo Life Sciences (±)Epibatidine (5mg). CAS: 140111-52-0
SDP
Supplier:  Enzo Life Sciences BMLC1140005
Encompass_Preferred
Epibatidine is an analgesic alkaloid isolated from the skin of the poison frog Epipedobates tricolor. It is the most potent nicotinic acetylcholine receptor agonist known. The natural (+) enantiomer is only 2-fold more potent than the (-) enantiomer with respect to analgesic activity in mice. Both enantiomers bind with high affinity (Ki=0.045 and 0.058 nM respectively) to rat brain membrane nicotinic receptors. Purity: ≥99% (HPLC). Solubility: Soluble in water (5 mg/ml) or DMSO (20 mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-0343