Learn More
Cayman Chemical (±)-Epibatidine (hydrochloride), Purity: ≥98%, CAS: 162885-01-0, 5 mg
SDP
Catalog No. 502989985
Encompass_Preferred
An agonist of α4β2 subunit-containing nAChRs (Ki = 43 pM); selective for α4β2 subunit-containing nAChRs over α7 nAChRs (Ki = 230 nM); 86Rb+ flux in IMR-32 cells (EC50 = 7 nM); induces dopamine release from rat striatal slices (EC50 = 0.4 nM); induces analgesia in the hot plate test and the Straub-tail response in mice (ED50s = 0.005 and 0.02 mg/kg, respectively)
Catalog No. 50-298-9985 Supplier Cayman Chemical Supplier No. 311935 MG
An agonist of α4β2 subunit-containing nAChRs (Ki = 43 pM); selective for α4β2 subunit-containing nAChRs over α7 nAChRs (Ki = 230 nM); 86Rb+ flux in IMR-32 cells (EC50 = 7 nM); induces dopamine release from rat striatal slices (EC50 = 0.4 nM); induces analgesia in the hot plate test and the Straub-tail response in mice (ED50s = 0.005 and 0.02 mg/kg, respectively)