Learn More
Cayman Chemical EpIberberIn 10mg
SDP
Catalog No. 502948693
Encompass_Preferred
An alkaloid with diverse biological activities; inhibits the activities of AChE, BChE, and BACE1 (IC50s = 1.07, 6.03, and 8.55 µM, respectively); scavenges peroxynitrite radicals in a cell-free assay (IC50 = 16.83 µM); is cytotoxic to HepG2 HCC cells at 100 µM; reduces serum total cholesterol and LDL-cholesterol levels in Syrian golden hamsters fed a high-fat and high-cholesterol diet at 70 mg/kg
Catalog No. 50-294-8693 Supplier Cayman Chemical Supplier No. 3456910 MG
An alkaloid with diverse biological activities; inhibits the activities of AChE, BChE, and BACE1 (IC50s = 1.07, 6.03, and 8.55 µM, respectively); scavenges peroxynitrite radicals in a cell-free assay (IC50 = 16.83 µM); is cytotoxic to HepG2 HCC cells at 100 µM; reduces serum total cholesterol and LDL-cholesterol levels in Syrian golden hamsters fed a high-fat and high-cholesterol diet at 70 mg/kg