Learn More
Medchemexpress LLC HY-N0226A 5mg Medchemexpress, Epiberberine (chloride) CAS:889665-86-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN0226A5MG
Encompass_Preferred
Medchemexpress, HY-N0226A 5mg Epiberberine (chloride) CAS:889665-86-5 Epiberberine chloride is an alkaloid isolated from Coptis chinensis, acts as a potent AChE and BChE inhibitor, and a non-competitive BACE1 inhibitor, with IC 50 s of 1.07, 6.03 and 8.55 μM, respectively. Epiberberine chloride has antioxidant activity, with peroxynitrite ONOO - scavenging effect (IC 50 , 16.83 μM), and may protect against Alzheimer disease [1] . Epiberberine chloride inhibits the early stage of differentiation of 3T3-L1 preadipocytes, downregulates the Raf/MEK1/2/ERK1/2 and AMPKα/Akt pathways [2] . Epiberberine has the potential effect in the research of diabetic disease [3] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3479