Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Epiberberine chloride | 889665-86-5 | MFCD01549686 | 99.0% | 371.81 | C20H18ClNO4 | 10 MG
SDP

Catalog No. 5000295669
Encompass_Preferred

Epiberberine chloride is a berberine-type alkaloid supplied for research use. It functions as a cholinesterase inhibitor and a non-competitive BACE1 inhibitor, and it exhibits antioxidant activity including peroxynitrite scavenging. The compound is provided as a solid for biochemical and pharmacological studies related to neurodegeneration and metabolic regulation.

  • Alkaloid with acetylcholinesterase and butyrylcholinesterase inhibitory activity.
  • Non-competitive BACE1 inhibitor, supporting amyloid-related research.
  • Antioxidant activity with ONOO- scavenging (IC50 16.83 μM).
  • Reported IC50s: AChE 1.07 μM, BChE 6.03 μM, BACE1 8.55 μM.
  • Molecular formula C20H18ClNO4; molecular weight 371.81.
  • Purity 99.02% (LCMS).
  • Storage: sealed at 4°C; in solution: -80°C (6 months), -20°C (1 month).
  • Available in small research pack sizes suitable for assay development and reference work.

Catalog No. 50-002-95669 Supplier Medchemexpress LLC Supplier No. HYN0226A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Epiberberine chloride is a berberine-type alkaloid supplied for research use. It functions as a cholinesterase inhibitor and a non-competitive BACE1 inhibitor, and it exhibits antioxidant activity including peroxynitrite scavenging. The compound is provided as a solid for biochemical and pharmacological studies related to neurodegeneration and metabolic regulation.

  • Alkaloid with acetylcholinesterase and butyrylcholinesterase inhibitory activity.
  • Non-competitive BACE1 inhibitor, supporting amyloid-related research.
  • Antioxidant activity with ONOO- scavenging (IC50 16.83 μM).
  • Reported IC50s: AChE 1.07 μM, BChE 6.03 μM, BACE1 8.55 μM.
  • Molecular formula C20H18ClNO4; molecular weight 371.81.
  • Purity 99.02% (LCMS).
  • Storage: sealed at 4°C; in solution: -80°C (6 months), -20°C (1 month).
  • Available in small research pack sizes suitable for assay development and reference work.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.