Learn More
TARGETMOL CHEMICALS INC EPIBRASSINOLIDE 25MG
SDP
Catalog No. 502576677
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Epibrassinolide (BP55) (EBR) is a biologically active compound of the brassinosteroids is a natural brassinosteroid (BR) derivative is a plant regulator with a similar structure to mammalian steroids. purity: 98%
Catalog No. 50-257-6677 Supplier TARGETMOL CHEMICALS INC Supplier No. T551425MG
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Epibrassinolide (BP55) (EBR) is a biologically active compound of the brassinosteroids is a natural brassinosteroid (BR) derivative is a plant regulator with a similar structure to mammalian steroids. purity: 98%