Learn More
Medchemexpress LLC HY-N0848 500mg , Epibrassinolide 24-Epibrassinolide;B1105;BP55 CAS:78821-43-9 Purity:98%
SDP
Catalog No. 501492368
Encompass_Preferred
Medchemexpress, HY-N0848 500mg Epibrassinolide 24-Epibrassinolide;B1105;BP55 CAS:78821-43-9 Formula:C28H48O6 Plant growth regulator, Apoptosis inducer Purity:98% Epibrassinolide is a natural brassinosteroid (BR) derivative, is a plant regulator with a similar structure to mammalian steroids. Epibrassinolide is a potential apoptotic inducer in various cancer cells without affecting the non-tumor cell growth. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2368 Supplier Medchemexpress LLC Supplier No. HYN0848500MG
Medchemexpress, HY-N0848 500mg Epibrassinolide 24-Epibrassinolide;B1105;BP55 CAS:78821-43-9 Formula:C28H48O6 Plant growth regulator, Apoptosis inducer Purity:98% Epibrassinolide is a natural brassinosteroid (BR) derivative, is a plant regulator with a similar structure to mammalian steroids. Epibrassinolide is a potential apoptotic inducer in various cancer cells without affecting the non-tumor cell growth. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.