Learn More
Tians Epic™ White Polypropylene Sleeves
Catalog No. 19138550 Shop All Tians International Products
Change view
Click to view available options
Cleanroom Class:
Class 10,000 (ISO 7)
Catalog No. Cleanroom Class
19-138-550 Class 10,000 (ISO 7)
1 options
This item is not returnable. View return policy
Catalog No. 19-138-550 Supplier Tians Supplier No. 8248818
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Cleanroom packed, low-lint sleeve for use in some critical environments and many general application areas.
  • Breathable non-woven polypropylene fabric
  • Latex free
  • Low linting
  • Elastic ends, serged seam construction
  • Tapered for proper fit
Specifications
Product Type Controlled Environment Sleeve
Construction Tapered
Sterility Non-sterile
Disposable Single-use
Closure Type Elastic Ends
Seam Type Serged
Latex Free Yes
Model 82488-18
Length (English) 18 in.
Length (Metric) 45.7 cm
Weight (Metric) 5 g
Cleanroom Class Class 10,000 (ISO 7)