Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tians Epic™ Heavy Polyethylene Shoe Covers

Catalog No. 19167175 Shop All Tians International Products
Change view
Click to view available options
Size:
Large
3X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-167-175 3X-Large
19-160-0526 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-167-175 Supplier Tians Supplier No. 747773XL
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Shoe covers constructed from a heavyweight polyethylene film for added strength and durability.

  • Lint-free, latex-free
  • Internally sewn elastic bottom seam
  • Heavyweight polyethylene film
  • Embossed texture for added skid resistance
  • For use in critical and low particulate clean environments and many general industrial applications
  • Fabric is suitable for exposure to bloodborne pathogens and some hazardous liquids
  • 6 mil material thickness
  • Suitable for ISO Cleanroom Class 5

Specifications

Size 3X-Large
Material Polyethylene
Color Rich Blue
Dimensions (L x W x H) 11.4 x 7 x 11.8 in. (2.89 x 17.78 x 29.97 cm)
Length (English) 11.4 in.
Length (Metric) 2.89 cm
Width (English) 7 in.
Width (Metric) 17.78 cm
Height (English) 11.8 in.
Height (Metric) 29.97 cm
Product Type Controlled Environments Shoe Covers
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.