Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tians Epic™ Shoe Cover

Catalog No. 19150324 Shop All Tians International Products
Change view
Click to view available options
Width (Metric):
12 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Width (Metric)
19-150-324 12 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-150-324 Supplier Tians Supplier No. 723883L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Clean and economical non-linting shoe cover for critical environments

  • Speeds up weighing process
  • Lint free disposable cleanroom shoe cover with heat sealed seams
  • Made from a durable soild polyethylene film
  • Fabric is constructed from a proprietary polyethylene blend, and is embossed for additional skid resistance
  • Seamless bottom
  • For use in critical and low particulate clean environments
  • Fabric is suitable for exposure to bloodborne pathogens, some hazardous liquids, and where liquid splash is a concern
  • Welded seams

Specifications

Size Universal
Material Polyethylene
Color White
Sterility Non-sterile
Closure Type Elastic Ankle
Model 723883-L
For Use With (Application) Cleanroom
Length (Metric) 40 cm
Width (Metric) 12 cm
Height (Metric) 15 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.