Learn More
TARGETMOL CHEMICALS INC Epicatechin 100MG
SDP
Catalog No. 502879273
Encompass_Preferred
Also available in 1 mL, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. (-)-Epicatechin (Epicatechin) is an inhibitor of cyclooxygenase-1 (COX-1), inhibiting the IL-1beta-induced expression of iNOS by blocking the nuclear localization of the p65 subunit of NF-kB. Purity 98%
Catalog No. 50-287-9273 Supplier TARGETMOL CHEMICALS INC Supplier No. T2856100MG
Also available in 1 mL, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. (-)-Epicatechin (Epicatechin) is an inhibitor of cyclooxygenase-1 (COX-1), inhibiting the IL-1beta-induced expression of iNOS by blocking the nuclear localization of the p65 subunit of NF-kB. Purity 98%