Learn More
Apexbio Technology LLC (-)-epicatechin 490-46-0 10mg
SDP
Catalog No. 5000063806
Encompass_Preferred
(-)-Epicatechin (CAS 490-46-0) is a flavonoid compound known for inducing pancreatic -cell regeneration and modulating nitric oxide (NO) metabolism It inhibits nitric oxide-related nitrite formation induced by interferon- and lipopolysaccharide in mouse macrophages and reduces peroxynitrite-mediated tyrosine nitration in HL-60 human leukemia cells Additionally in human endothelial cells (HUVECs) it enhances NO production displaying vasodilatory properties In diabetic rat models (alloxan-induced) (-)-epicatechin administration promotes -cell recovery reduces hyperglycemia and prevents -cell necrosis This profile underscores its research relevance for diabetes and vascular biology investigations
Catalog No. 50-000-63806 Supplier Apexbio Technology LLC Supplier No. B496410
(-)-Epicatechin (CAS 490-46-0) is a flavonoid compound known for inducing pancreatic -cell regeneration and modulating nitric oxide (NO) metabolism It inhibits nitric oxide-related nitrite formation induced by interferon- and lipopolysaccharide in mouse macrophages and reduces peroxynitrite-mediated tyrosine nitration in HL-60 human leukemia cells Additionally in human endothelial cells (HUVECs) it enhances NO production displaying vasodilatory properties In diabetic rat models (alloxan-induced) (-)-epicatechin administration promotes -cell recovery reduces hyperglycemia and prevents -cell necrosis This profile underscores its research relevance for diabetes and vascular biology investigations