Learn More
Medchemexpress LLC HY-N0001 100mg , (-)-Epicatechin (-)-Epicatechol;Epicatechin;epi-Catechin CAS:490-46-0 Purity:98%
SDP
Catalog No. 501493495
Encompass_Preferred
Medchemexpress, HY-N0001 100mg (-)-Epicatechin (-)-Epicatechol;Epicatechin;epi-Catechin CAS:490-46-0 Formula:C15H14O6 IC50: 3.2 μM (COX-1) Purity:98% (-)-Epicatechin inhibits cyclooxygenase-1 ( COX-1 ) with an IC 50 of 3.2 μM. (-)-Epicatechin inhibits the IL-1β-induced expression of iNOS by blocking the nuclear localization of the p65 subunit of NF-κB. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-3495 Supplier Medchemexpress LLC Supplier No. HYN0001100MG
Medchemexpress, HY-N0001 100mg (-)-Epicatechin (-)-Epicatechol;Epicatechin;epi-Catechin CAS:490-46-0 Formula:C15H14O6 IC50: 3.2 μM (COX-1) Purity:98% (-)-Epicatechin inhibits cyclooxygenase-1 ( COX-1 ) with an IC 50 of 3.2 μM. (-)-Epicatechin inhibits the IL-1β-induced expression of iNOS by blocking the nuclear localization of the p65 subunit of NF-κB. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.