Learn More
Selleck Chemical LLC (-)Epicatechin S4723-25mg
SDP
Catalog No. 503109451
Encompass_Preferred
( )Epicatechin (L-Epicatechin (-)-Epicatechol) is a flavonoid present in cocoa green tea and red wine It is a strong antioxidant has insulin mimic action and improves heart health
Catalog No. 50-310-9451 Supplier Selleck Chemical LLC Supplier No. S472325MG
( )Epicatechin (L-Epicatechin (-)-Epicatechol) is a flavonoid present in cocoa green tea and red wine It is a strong antioxidant has insulin mimic action and improves heart health