Learn More
Medchemexpress LLC (-)-Epicatechin | 490-46-0 | 95.6% | 290.27 | 1 MG
SDP
Catalog No. 5000428648
Encompass_Preferred
(-)-Epicatechin is the analytical standard of (-)-Epicatechin, intended for research and analytical applications. It inhibits cyclooxygenase-1 (COX-1) and the IL-1β-induced expression of iNOS by blocking the nuclear localization of the p65 subunit of NF-κB. This analytical standard is commonly used in qualitative, quantitative, and methodological research experiments in techniques such as HPLC, GC, and MS.
  • Suitable for research and analytical applications.
  • Inhibits cyclooxygenase-1.
  • Blocks nuclear localization of p65 subunit of NF-κB.
  • Used in qualitative research experiments.
  • Used in quantitative research experiments.
  • Used in methodological research experiments.
  • Compatible with HPLC, GC, and MS techniques.
Catalog No. 50-004-28648 Supplier Medchemexpress LLC Supplier No. HYN0001R1MG
(-)-Epicatechin is the analytical standard of (-)-Epicatechin, intended for research and analytical applications. It inhibits cyclooxygenase-1 (COX-1) and the IL-1β-induced expression of iNOS by blocking the nuclear localization of the p65 subunit of NF-κB. This analytical standard is commonly used in qualitative, quantitative, and methodological research experiments in techniques such as HPLC, GC, and MS.
  • Suitable for research and analytical applications.
  • Inhibits cyclooxygenase-1.
  • Blocks nuclear localization of p65 subunit of NF-κB.
  • Used in qualitative research experiments.
  • Used in quantitative research experiments.
  • Used in methodological research experiments.
  • Compatible with HPLC, GC, and MS techniques.