Learn More
Medchemexpress LLC (+)-epicatechin (dexepicatechin) | 35323-91-2 | MFCD00075648 | 99.6% | 10 MG
SDP
Catalog No. 5000297580
Encompass_Preferred
(+)-Epicatechin is a chiral catechin and polyphenolic antioxidant supplied as a research reagent. It is commonly used in biochemical and analytical assays to evaluate antioxidant activity and enzyme inhibition. The compound is a white to off-white solid with molecular formula C15H14O6 and a molecular weight of 290.27 g/mol. Purity is typically reported at about 99.6%.
  • High purity suitable for analytical applications
  • Solid form convenient for storage and handling
  • Used in antioxidant and cyclooxygenase inhibition studies
  • Soluble in common organic solvents for stock solutions
  • Provided in a small quantity suitable for early-stage experiments
Catalog No. 50-002-97580 Supplier Medchemexpress LLC Supplier No. HYN0001A10MG
(+)-Epicatechin is a chiral catechin and polyphenolic antioxidant supplied as a research reagent. It is commonly used in biochemical and analytical assays to evaluate antioxidant activity and enzyme inhibition. The compound is a white to off-white solid with molecular formula C15H14O6 and a molecular weight of 290.27 g/mol. Purity is typically reported at about 99.6%.
  • High purity suitable for analytical applications
  • Solid form convenient for storage and handling
  • Used in antioxidant and cyclooxygenase inhibition studies
  • Soluble in common organic solvents for stock solutions
  • Provided in a small quantity suitable for early-stage experiments