Learn More
Abcam Epicatechin, Flavonoid, 1MG
Catalog No. 502706886
Encompass_Preferred
MW 290.27 Da, Purity >99%. Flavonoid. Displays multiple biological effects including antioxidant, antinociceptive, cardioprotective, anti-inflammatory and antineoplastic effects.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6886 Supplier Abcam Supplier No. AB1411191MG
MW 290.27 Da, Purity >99%. Flavonoid. Displays multiple biological effects including antioxidant, antinociceptive, cardioprotective, anti-inflammatory and antineoplastic effects.
The product is subject to the following: Abcam Restricted Use Statement