Learn More
Cayman Chemical (-)-Epicatechin Gallate, Purity: ≥98%, CAS: 1257-08-5, 10 mg
SDP
Catalog No. 502951998
Encompass_Preferred
A natural catechin which inhibits the growth of cancer cells and has anti-inflammatory effects; has potent antioxidant activity and facilitates the killing of methicillin-resistant strains of S. aureus
Catalog No. 50-295-1998 Supplier Cayman Chemical Supplier No. 1180810 MG
A natural catechin which inhibits the growth of cancer cells and has anti-inflammatory effects; has potent antioxidant activity and facilitates the killing of methicillin-resistant strains of S. aureus