Learn More
Cayman Chemical EpIcatechIn Galate 25mg
SDP
Catalog No. 502951961
Encompass_Preferred
A natural catechin which inhibits the growth of cancer cells and has anti-inflammatory effects; has potent antioxidant activity and facilitates the killing of methicillin-resistant strains of S. aureus
Catalog No. 50-295-1961 Supplier Cayman Chemical Supplier No. 1180825 MG
A natural catechin which inhibits the growth of cancer cells and has anti-inflammatory effects; has potent antioxidant activity and facilitates the killing of methicillin-resistant strains of S. aureus