Learn More
Medchemexpress LLC (-)-Epicatechin gallate | 1257-08-5 | MFCD00075936 | 99.6% | 442.37 | C22H18O10 | 10 MG
SDP
Catalog No. 5000298986
Encompass_Preferred
(-)-Epicatechin gallate is a flavonoid gallate used as a research reagent for biochemical and cell-based studies. It inhibits cyclooxygenase-1 (COX-1) with a reported IC50 of approximately 7.5 μM and has reported activity against glucose transporters (GLUT1, GLUT5). The compound is provided as a high-purity analytical reagent suitable for assay development and mechanistic studies.
  • Natural flavonoid derivative
  • Inhibits COX-1 (IC50 ≈ 7.5 μM)
  • Reported GLUT1 and GLUT5 inhibitor
  • Molecular formula C22H18O10, molecular weight 442.37
  • High purity (HPLC) - 99.6%
  • Supplied in small research pack sizes for analytical use
Catalog No. 50-002-98986 Supplier Medchemexpress LLC Supplier No. HYN000210MG
(-)-Epicatechin gallate is a flavonoid gallate used as a research reagent for biochemical and cell-based studies. It inhibits cyclooxygenase-1 (COX-1) with a reported IC50 of approximately 7.5 μM and has reported activity against glucose transporters (GLUT1, GLUT5). The compound is provided as a high-purity analytical reagent suitable for assay development and mechanistic studies.
  • Natural flavonoid derivative
  • Inhibits COX-1 (IC50 ≈ 7.5 μM)
  • Reported GLUT1 and GLUT5 inhibitor
  • Molecular formula C22H18O10, molecular weight 442.37
  • High purity (HPLC) - 99.6%
  • Supplied in small research pack sizes for analytical use