Learn More
Medchemexpress LLC (-)-Epicatechin gallate (standard) | 1257-08-5 | 442.37 | 1 MG
SDP
Catalog No. 5000428655
Encompass_Preferred
(-)-Epicatechin gallate (Standard) is the analytical standard of (-)-Epicatechin gallate. This product is intended for research and analytical applications. It inhibits cyclooxygenase-1 (COX-1) with an IC50 of 7.5 μM, and is a GLUT1 and GLUT5 inhibitor, decreasing cell growth of GLUT5-expressing hxt 0 yeast cells.
  • Targets: COX, autophagy, virus protease, GLUT
  • Research areas: Infection, cancer
  • Structural classification: Flavonoids, flavanols, phenols, polyphenols
  • Source classification: *Camellia sinensis* (L.) O. Ktze. plants, Theaceae
Catalog No. 50-004-28655 Supplier Medchemexpress LLC Supplier No. HYN0002R1MG
(-)-Epicatechin gallate (Standard) is the analytical standard of (-)-Epicatechin gallate. This product is intended for research and analytical applications. It inhibits cyclooxygenase-1 (COX-1) with an IC50 of 7.5 μM, and is a GLUT1 and GLUT5 inhibitor, decreasing cell growth of GLUT5-expressing hxt 0 yeast cells.
  • Targets: COX, autophagy, virus protease, GLUT
  • Research areas: Infection, cancer
  • Structural classification: Flavonoids, flavanols, phenols, polyphenols
  • Source classification: *Camellia sinensis* (L.) O. Ktze. plants, Theaceae