Learn More
Medchemexpress LLC (-)-Epicatechin gallate (Standard) | 1257-08-5 | 97.7% | 442.37 | 50 MG
SDP
Catalog No. 5000428656
Encompass_Preferred
(-)-Epicatechin gallate (Standard) is the analytical standard of (-)-Epicatechin gallate. This product is intended for research and analytical applications. It inhibits cyclooxygenase-1 (COX-1) with an IC50 of 7.5 μM. It is also a GLUT1 and GLUT5 inhibitor, and decreases cell growth of GLUT5-expressing hxt0 yeast cells.
  • Analytical standard for (-)-Epicatechin gallate.
  • Intended for research and analytical applications.
  • Inhibits cyclooxygenase-1 (COX-1).
  • Acts as a GLUT1 and GLUT5 inhibitor.
  • Decreases cell growth of GLUT5-expressing hxt0 yeast cells.
  • Commonly used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS.
Catalog No. 50-004-28656 Supplier Medchemexpress LLC Supplier No. HYN0002R50MG
(-)-Epicatechin gallate (Standard) is the analytical standard of (-)-Epicatechin gallate. This product is intended for research and analytical applications. It inhibits cyclooxygenase-1 (COX-1) with an IC50 of 7.5 μM. It is also a GLUT1 and GLUT5 inhibitor, and decreases cell growth of GLUT5-expressing hxt0 yeast cells.
  • Analytical standard for (-)-Epicatechin gallate.
  • Intended for research and analytical applications.
  • Inhibits cyclooxygenase-1 (COX-1).
  • Acts as a GLUT1 and GLUT5 inhibitor.
  • Decreases cell growth of GLUT5-expressing hxt0 yeast cells.
  • Commonly used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS.