Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (-)-epicatechin (standard) | 490-46-0 | 290.27 g/mol | C15H14O6 | 10 MG
SDP

Catalog No. 5000297556
Encompass_Preferred

(-)-Epicatechin (standard) is an analytical reference material of the flavonoid (-)-epicatechin provided as a 10 mg quantity for research and analytical use. It is intended for assay calibration, method development, and reference measurements; not for human or clinical use. Recommended storage is 4°C sealed and protected from light and moisture; in solvent, store at -80°C for up to 6 months or -20°C for up to 1 month.

  • Analytical reference standard of (-)-epicatechin.
  • Intended for research and analytical applications; not for human use.
  • CAS number 490-46-0; molecular formula C15H14O6.
  • Supplied as a 10 mg quantity suitable for calibration and method development.
  • Recommended storage: 4°C sealed; in solvent store at -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-97556 Supplier Medchemexpress LLC Supplier No. HYN0001R10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

(-)-Epicatechin (standard) is an analytical reference material of the flavonoid (-)-epicatechin provided as a 10 mg quantity for research and analytical use. It is intended for assay calibration, method development, and reference measurements; not for human or clinical use. Recommended storage is 4°C sealed and protected from light and moisture; in solvent, store at -80°C for up to 6 months or -20°C for up to 1 month.

  • Analytical reference standard of (-)-epicatechin.
  • Intended for research and analytical applications; not for human use.
  • CAS number 490-46-0; molecular formula C15H14O6.
  • Supplied as a 10 mg quantity suitable for calibration and method development.
  • Recommended storage: 4°C sealed; in solvent store at -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.