Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (+/-)-Epichlorohydrin

Catalog No. 502897862
Encompass
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-289-7862 2.5 L
50-289-8640 1 L
50-184-4879 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 50-289-7862 Supplier Sigma Aldrich Supplier No. 453402.5LF
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(+/-)-Epichlorohydrin
TRUSTED_SUSTAINABILITY

Specifications

CAS 106-89-8
Melting Point -57°C (lit.)
Density 1.183 g/mL (at 25°C (literature))
Boiling Point 115°C to 117°C (lit.)
Molecular Formula C3H5ClO
Refractive Index n20/D 1.438 (literature); n20/D 1.438
Linear Formula C3H5ClO
MDL Number MFCD00005132
Quantity 2.5 L
Synonym (+/-)-2-(Chloromethyl)oxirane; 1-Chloro-2,3-epoxypropane
Molecular Weight (g/mol) 92.52
Percent Purity ≥99% (GC)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.