Learn More
STEMCELL Technologies EpiCult™ Plus Medium, Size: 500 mL
SDP
Catalog No. 501974375
Encompass
EpiCult™ Plus Medium is a serum- and BPE-free culture medium for the expansion of epithelial stem cells from mammary, prostate, and other stratified and pseudostratified epithelial tissues from human, mouse, and rat. Epithelial cells can be expanded in EpiCult™ Plus Medium for multiple passages and for > 30 population doublings; for some human mammary cells, addition of fetal bovine serum may be required. EpiCult™ Plus Medium is not recommended for human keratinocytes or primary human airway cells; for culturing primary human airway cells, we recommend PneumaCult™-Ex Plus Medium (Catalog #05040). Note: Hydrocortisone Stock Solution (Catalog #07925) is required for preparation of complete EpiCult™ Plus Medium.
Catalog No. 50-197-4375 Supplier STEMCELL Technologies Supplier No. 06070
EpiCult™ Plus Medium is a serum- and BPE-free culture medium for the expansion of epithelial stem cells from mammary, prostate, and other stratified and pseudostratified epithelial tissues from human, mouse, and rat. Epithelial cells can be expanded in EpiCult™ Plus Medium for multiple passages and for > 30 population doublings; for some human mammary cells, addition of fetal bovine serum may be required. EpiCult™ Plus Medium is not recommended for human keratinocytes or primary human airway cells; for culturing primary human airway cells, we recommend PneumaCult™-Ex Plus Medium (Catalog #05040). Note: Hydrocortisone Stock Solution (Catalog #07925) is required for preparation of complete EpiCult™ Plus Medium.