Learn More
Zen Bio Inc EPID KERATINOCYTE/DOG MEDIUM
SDP
Catalog No. NC9761642
Encompass
Non-defined media optimized for the growth of sheep and dog keratinocytes
Catalog No. NC9761642 Supplier Zen Bio Inc Supplier No. CNT09
Non-defined media optimized for the growth of sheep and dog keratinocytes