Learn More
TARGETMOL CHEMICALS INC EPIDERMAL GROWTH FACTOR 5MG
SDP
Catalog No. 502579053
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Epidermal Growth Factor binds to epidermal growth factor receptor (EGFR) and stimulates cell growth and proliferation. It is approved for the treatment of diabetic foot ulcers. purity: 100%
Catalog No. 50-257-9053 Supplier TARGETMOL CHEMICALS INC Supplier No. T86795MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Epidermal Growth Factor binds to epidermal growth factor receptor (EGFR) and stimulates cell growth and proliferation. It is approved for the treatment of diabetic foot ulcers. purity: 100%