Learn More
Cusabio Technology LLC Epidermal growth factor EGF
Catalog No. 5000049084
Encompass_Preferred
The Mouse Epidermal Growth Factor (EGF) ELISA Kit is designed to detect and quantify EGF levels in serum plasma and tissue homogenates with high precision This kit employs a sandwich ELISA technique offering a broad detection range of 7 8 pg/mL to 500 pg/mL and a sensitivity of 1 95 pg/mL ensuring accurate measurement of low-concentration samples The assay requires a sample volume of 50-100 L and typically delivers results within 1-5 hours optimized for efficiency in research settings Quantitative analysis is performed by measuring absorbance at 450 nm providing reliable data for studies involving EGF-related cellular processes wound healing or tissue regeneration Its standardized protocol ensures reproducibility across experimental conditions making it suitable for both basic research and preclinical applications
Catalog No. 50-000-49084 Supplier Cusabio Technology LLC Supplier No. CSBE08028M
The Mouse Epidermal Growth Factor (EGF) ELISA Kit is designed to detect and quantify EGF levels in serum plasma and tissue homogenates with high precision This kit employs a sandwich ELISA technique offering a broad detection range of 7 8 pg/mL to 500 pg/mL and a sensitivity of 1 95 pg/mL ensuring accurate measurement of low-concentration samples The assay requires a sample volume of 50-100 L and typically delivers results within 1-5 hours optimized for efficiency in research settings Quantitative analysis is performed by measuring absorbance at 450 nm providing reliable data for studies involving EGF-related cellular processes wound healing or tissue regeneration Its standardized protocol ensures reproducibility across experimental conditions making it suitable for both basic research and preclinical applications