Learn More
Description
- Epidermal Growth Factor, Human, Recombinant is a recombinant analog of human epidermal growth factor (EGF) that provides a potent and cost-effective mitogen for cell culture applications.
- It is ideal for both research and large-scale culture systems utilizing serum-free or low-level serum applications.
- It is effective in a variety of fibroblast and endothelial cell types and produces extremely potent results in cell culture.
- It is produced in a patented E. coli expression system free of animal-derived components.
- Sterile filtered and then the protein was lyophilized with no additives.
Specifications
Specifications
| CAS | 62229-50-9 |
| Concentration | 0.5 to 25.0 ng/mL |
| Form | Lyophilized |
| Purity or Quality Grade | ≥98% |
| Quantity | 100 μg |
Safety and Handling
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.