Learn More
CPC Scientific H-Asp-Val-Val-Asp-Ala-Asp-Glu-Tyr-Leu-Ile-Pro-Gln-OH (trifluoroacetate salt) 0.5MG
SDP
Catalog No. 502109394
Encompass
EGFR fragment that contains the autophosphorylation site at Tyr992.ONE-LETTER SEQUENCE: DVVDADEYLIPQMOLECULAR FORMULA: C61H93N13O23MOLECULAR WEIGHT:1376.5STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [96249-43-3], RESEARCH AREA: CancerREFERENCES: A. Ullrich et al., Nature, 309, 418 (1984)
Catalog No. 50-210-9394 Supplier CPC Scientific Supplier No. GRFF002A
EGFR fragment that contains the autophosphorylation site at Tyr992.ONE-LETTER SEQUENCE: DVVDADEYLIPQMOLECULAR FORMULA: C61H93N13O23MOLECULAR WEIGHT:1376.5STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [96249-43-3], RESEARCH AREA: CancerREFERENCES: A. Ullrich et al., Nature, 309, 418 (1984)