Learn More
Apexbio Technology LLC Epidermal growth factor receptor (994-1002) acetyl/amide 1mg
SDP
Catalog No. 5000097104
Encompass_Preferred
Epidermal growth factor receptor 994-1002 acetyl amide is a peptide fragment derived from the epidermal growth factor receptor (EGFR) acetylated and amidated at its termini It is designed to serve as a tool for investigating receptor-ligand interactions and receptor activation mechanisms associated with EGFR-mediated signaling pathways Epidermal growth factor receptor 994-1002 acetyl amide acts by enabling the study of EGFR activation particularly receptor dimerization and autophosphorylation processes Based on its biological characteristics this peptide has research potential in evaluating therapeutic strategies targeting EGFR-mediated oncogenesis including studies of proliferation differentiation migration and survival in malignancies such as glioblastoma and lung cancers
Catalog No. 50-000-97104 Supplier Apexbio Technology LLC Supplier No. A11331
Epidermal growth factor receptor 994-1002 acetyl amide is a peptide fragment derived from the epidermal growth factor receptor (EGFR) acetylated and amidated at its termini It is designed to serve as a tool for investigating receptor-ligand interactions and receptor activation mechanisms associated with EGFR-mediated signaling pathways Epidermal growth factor receptor 994-1002 acetyl amide acts by enabling the study of EGFR activation particularly receptor dimerization and autophosphorylation processes Based on its biological characteristics this peptide has research potential in evaluating therapeutic strategies targeting EGFR-mediated oncogenesis including studies of proliferation differentiation migration and survival in malignancies such as glioblastoma and lung cancers