Learn More
EPICYPHER INC EPIDYNE-FRET NUCLEOSOME REMODE
SDP
Catalog No. NC1417675
Encompass
EpiDyne-FRET Nucleosome Remodeling Assay Substrate 50ugMononucleosomes assembled from recombinant human histones expressed in E. coli
Catalog No. NC1417675 Supplier EPICYPHER INC Supplier No. 164201
EpiDyne-FRET Nucleosome Remodeling Assay Substrate 50ugMononucleosomes assembled from recombinant human histones expressed in E. coli