Learn More
AdipoGen Epifluorohydrin
SDP
Supplier:  AdipoGen CDXE0243G001
Encompass_Preferred
Chemical. CAS 503-09-3. Formula C3H5FO. Molecular Weight 76.07. Epifluorohydrin is used as an organic building block for chemical synthesis.
Catalog No. 50-256-0961