Learn More
Medchemexpress LLC Epifriedelanol | 16844-71-6 | 428.73 | 5 MG
SDP
Catalog No. 5000415046
Encompass_Preferred
Epifriedelanol is a triterpenoid isolated from the root bark of Ulmus davidiana, known for its antitumor and anti-inflammatory activities. It induces apoptosis in DU145 cells and inhibits cellular senescence in human primary cells. Research indicates it can counteract Bisphenol A (BPA)-induced gonadotoxicity and reduce peroxidation while suppressing inflammatory markers.
  • Triterpenoid compound.
  • Induces apoptosis in DU145 cells.
  • Exhibits antitumor and anti-inflammatory activities.
  • Inhibits cellular senescence.
  • Mitigates Bisphenol A (BPA)-induced gonadotoxicity.
  • Supports biosafety in vivo.
  • Reduces peroxidation and suppresses inflammatory markers.
Catalog No. 50-004-15046 Supplier Medchemexpress LLC Supplier No. HYN19315MG
Epifriedelanol is a triterpenoid isolated from the root bark of Ulmus davidiana, known for its antitumor and anti-inflammatory activities. It induces apoptosis in DU145 cells and inhibits cellular senescence in human primary cells. Research indicates it can counteract Bisphenol A (BPA)-induced gonadotoxicity and reduce peroxidation while suppressing inflammatory markers.
  • Triterpenoid compound.
  • Induces apoptosis in DU145 cells.
  • Exhibits antitumor and anti-inflammatory activities.
  • Inhibits cellular senescence.
  • Mitigates Bisphenol A (BPA)-induced gonadotoxicity.
  • Supports biosafety in vivo.
  • Reduces peroxidation and suppresses inflammatory markers.