Learn More
Medchemexpress LLC Epifriedelanol | 16844-71-6 | 99.7% | 428.73 g/mol | C30H52O | 10 MG
SDP
Catalog No. 5000295604
Encompass_Preferred
Epifriedelanol is a triterpenoid research compound isolated from the root bark of Ulmus davidiana. It has reported antitumor and anti-inflammatory activity, and induces apoptosis in DU145 cells; supplied for research use only.
  • High purity compound suitable for biochemical and cellular studies.
  • Reported activities include apoptosis induction and anti-inflammatory effects.
  • Recommended storage: 4°C, protect from light; in solvent: -80°C (6 months) or -20°C (1 month).
  • Provided as a 10 mg research pack for laboratory assays.
Catalog No. 50-002-95604 Supplier Medchemexpress LLC Supplier No. HYN193110MG
Epifriedelanol is a triterpenoid research compound isolated from the root bark of Ulmus davidiana. It has reported antitumor and anti-inflammatory activity, and induces apoptosis in DU145 cells; supplied for research use only.
  • High purity compound suitable for biochemical and cellular studies.
  • Reported activities include apoptosis induction and anti-inflammatory effects.
  • Recommended storage: 4°C, protect from light; in solvent: -80°C (6 months) or -20°C (1 month).
  • Provided as a 10 mg research pack for laboratory assays.