Learn More
Cayman Chemical EpIfrIedelnol 10mg
SDP
Catalog No. 502912573
Encompass_Preferred
A triterpene with anticancer and senescence modifying activities; increases levels of P-gp and MDR2 and enhances doxorubicin-induced cytotoxicity in doxorubicin-resistant K562/ADM cells from 5-20 µM; inhibits doxorubicin-induced senescence in isolated HDFs, as well as replicative senescence in HDFs and HUVECs
Catalog No. 50-291-2573 Supplier Cayman Chemical Supplier No. 3614810 MG
A triterpene with anticancer and senescence modifying activities; increases levels of P-gp and MDR2 and enhances doxorubicin-induced cytotoxicity in doxorubicin-resistant K562/ADM cells from 5-20 µM; inhibits doxorubicin-induced senescence in isolated HDFs, as well as replicative senescence in HDFs and HUVECs