Learn More
Medchemexpress LLC HY-N2228 1mg Medchemexpress, (-)-Epigallocatechin-3-(3''-O-methyl) gallate CAS:83104-87-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN22281MG
Encompass_Preferred
Medchemexpress, HY-N2228 1mg (-)-Epigallocatechin-3-(3''-O-methyl) gallate CAS:83104-87-4 (-)-Epigallocatechin-3-(3''-O-methyl) gallate is a natural product isolated from the tea leaf, with strong antioxidative activity. (-)-Epigallocatechin-3-(3''-O-methyl) gallate has a strong cytotoxic activity for rat cancer cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0376