Learn More
Medchemexpress LLC (-)-Epigallocatechin-3-(3''-O-methyl) gallate | 83104-87-4 | 5 MG
SDP
Catalog No. 5000420525
Encompass_Preferred
(-)-Epigallocatechin-3-(3''-O-methyl) gallate ((-)-EGCG-3''-O-ME) is a natural product isolated from tea leaf with strong antioxidative activity. It also exhibits potent cytotoxic activity for rat cancer cells.
  • Isolated from tea leaves.
  • Exhibits strong antioxidative activity.
  • Shows potent cytotoxic activity against rat cancer cells.
  • Classified as a flavonoid, flavanol, phenol, and polyphenol.
  • Scavenges DPPH-derived and ABTS radicals.
  • Scavenges intracellular ROS production.
  • Promotes cell proliferation.
Catalog No. 50-004-20525 Supplier Medchemexpress LLC Supplier No. HYN22285MG
(-)-Epigallocatechin-3-(3''-O-methyl) gallate ((-)-EGCG-3''-O-ME) is a natural product isolated from tea leaf with strong antioxidative activity. It also exhibits potent cytotoxic activity for rat cancer cells.
  • Isolated from tea leaves.
  • Exhibits strong antioxidative activity.
  • Shows potent cytotoxic activity against rat cancer cells.
  • Classified as a flavonoid, flavanol, phenol, and polyphenol.
  • Scavenges DPPH-derived and ABTS radicals.
  • Scavenges intracellular ROS production.
  • Promotes cell proliferation.