Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (-)-Epigallocatechin-3-(3''-O-methyl) gallate | 83104-87-4 | MFCD06799452 | 99.8% | 472.40 g/mol | C23H20O11 | 10 MG
SDP

Catalog No. 5000296455
Encompass_Preferred

(-)-Epigallocatechin-3-(3''-O-methyl) gallate is a methylated epigallocatechin derivative commonly used in biochemical and pharmacological research to study antioxidant and anticancer activities. It is provided in small quantities for screening, analytical, and cellular assays.

  • High purity (HPLC 99.8%)
  • Molecular weight 472.40 g/mol
  • Chemical formula C23H20O11
  • CAS number 83104-87-4
  • Supplied as a 10 mg research reagent suitable for assay development and compound screening

Catalog No. 50-002-96455 Supplier Medchemexpress LLC Supplier No. HYN222810MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(-)-Epigallocatechin-3-(3''-O-methyl) gallate is a methylated epigallocatechin derivative commonly used in biochemical and pharmacological research to study antioxidant and anticancer activities. It is provided in small quantities for screening, analytical, and cellular assays.

  • High purity (HPLC 99.8%)
  • Molecular weight 472.40 g/mol
  • Chemical formula C23H20O11
  • CAS number 83104-87-4
  • Supplied as a 10 mg research reagent suitable for assay development and compound screening

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.